accessibility menu, dialog, popup

UserName

CAS RN 1155264-46-2

IUPAC Name: 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine
Synonyms: 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine
Molecular Weight (g/mol): 261.13
Molecular Formula: C14H20BNO3
InChi Key: HFUHUNYUUCDCAU-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(OCCN2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results