missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dihydro-2H-1,4-benzoxazine-2-carbonitrile, 97+%, Thermo Scientific™
$176.38 - $176.38
Chemical Identifiers
| CAS | 86267-86-9 |
|---|---|
| Molecular Formula | C9H8N2O |
| Molecular Weight (g/mol) | 160.176 |
| InChI Key | YSTANLOUKDVPGJ-UHFFFAOYSA-N |
| Synonym | 3,4-dihydro-2h-benzo b 1,4 oxazine-2-carbonitrile, 2h,3h,4h-benzo e 1,4-oxazaperhydroine-2-carbonitrile, 2h-1,4-benzoxazine-2-carbonitrile,3,4-dihydro |
| PubChem CID | 2795504 |
| IUPAC Name | 3,4-dihydro-2H-1,4-benzoxazine-2-carbonitrile |
| SMILES | C1C(OC2=CC=CC=C2N1)C#N |
Chemical Identifiers
| 86267-86-9 | |
| 160.176 | |
| 3,4-dihydro-2h-benzo b 1,4 oxazine-2-carbonitrile, 2h,3h,4h-benzo e 1,4-oxazaperhydroine-2-carbonitrile, 2h-1,4-benzoxazine-2-carbonitrile,3,4-dihydro | |
| 3,4-dihydro-2H-1,4-benzoxazine-2-carbonitrile |
| C9H8N2O | |
| YSTANLOUKDVPGJ-UHFFFAOYSA-N | |
| 2795504 | |
| C1C(OC2=CC=CC=C2N1)C#N |
Specifications
| 86267-86-9 | |
| Amber Glass Bottle | |
| 1 g | |
| YSTANLOUKDVPGJ-UHFFFAOYSA-N | |
| 3,4-dihydro-2H-1,4-benzoxazine-2-carbonitrile | |
| 2795504 | |
| 97% |
| 97% | |
| C9H8N2O | |
| 3,4-dihydro-2h-benzo b 1,4 oxazine-2-carbonitrile, 2h,3h,4h-benzo e 1,4-oxazaperhydroine-2-carbonitrile, 2h-1,4-benzoxazine-2-carbonitrile,3,4-dihydro | |
| C1C(OC2=CC=CC=C2N1)C#N | |
| 160.176 | |
| 160.18 | |
| 3, 4-Dihydro-2H-1, 4-benzoxazine-2-carbonitrile |