Learn More
3,4-Dihydro-2H-1,4-benzothiazine, 97%
CAS: 3080-99-7 | C8H9NS | 151.23 g/mol
Supplier: Thermo Scientific Chemicals H6104503
Description
3,4-Dihydro-2H-1,4-benzothiazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Dihydro-2H-1,4-benzothiazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| 3,4-Dihydro-2H-1,4-benzothiazine | |
| C8H9NS | |
| 1 g | |
| YBBLSBDJIKMXNQ-UHFFFAOYSA-N | |
| 3,4-dihydro-2H-1,4-benzothiazine | |
| 151064 | |
| 97% |
| 3080-99-7 | |
| MFCD04038593 | |
| 3,4-dihydro-2h-benzo b 1,4 thiazine, 2,3-dihydro-1,4-benzothiazine, 3,4-dihydro-2h-benzo 1,4 thiazine, 2h-1,4-benzothiazine, 3,4-dihydro, benzothiomorpholine, thiaphenomorpholine, 2,3-dihydrobenzo-1,4-thiazine, 2,3-dihydro-4h-1,4-benzothiazine, 2h,3h,4h-benzo e 1,4-thiazine, 2h-1,4-benzothiazine,3,4-dihydro | |
| C1CSC2=CC=CC=C2N1 | |
| 151.23 | |
| 151.23 |
Chemical Identifiers
| 3080-99-7 | |
| 151.23 | |
| YBBLSBDJIKMXNQ-UHFFFAOYSA-N | |
| 151064 | |
| C1CSC2=CC=CC=C2N1 |
| C8H9NS | |
| MFCD04038593 | |
| 3,4-dihydro-2h-benzo b 1,4 thiazine, 2,3-dihydro-1,4-benzothiazine, 3,4-dihydro-2h-benzo 1,4 thiazine, 2h-1,4-benzothiazine, 3,4-dihydro, benzothiomorpholine, thiaphenomorpholine, 2,3-dihydrobenzo-1,4-thiazine, 2,3-dihydro-4h-1,4-benzothiazine, 2h,3h,4h-benzo e 1,4-thiazine, 2h-1,4-benzothiazine,3,4-dihydro | |
| 3,4-dihydro-2H-1,4-benzothiazine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only