missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,4-benzothiazine, 97%

Catalog No. p-7041097
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6104503 1 g
AAH6104506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6104503 Supplier Thermo Scientific Chemicals Supplier No. H6104503

CAS: 3080-99-7 | C8H9NS | 151.23 g/mol

3,4-Dihydro-2H-1,4-benzothiazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydro-2H-1,4-benzothiazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 3080-99-7
Molecular Formula C8H9NS
Molecular Weight (g/mol) 151.23
MDL Number MFCD04038593
InChI Key YBBLSBDJIKMXNQ-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-benzo b 1,4 thiazine, 2,3-dihydro-1,4-benzothiazine, 3,4-dihydro-2h-benzo 1,4 thiazine, 2h-1,4-benzothiazine, 3,4-dihydro, benzothiomorpholine, thiaphenomorpholine, 2,3-dihydrobenzo-1,4-thiazine, 2,3-dihydro-4h-1,4-benzothiazine, 2h,3h,4h-benzo e 1,4-thiazine, 2h-1,4-benzothiazine,3,4-dihydro
PubChem CID 151064
IUPAC Name 3,4-dihydro-2H-1,4-benzothiazine
SMILES C1CSC2=CC=CC=C2N1

Specifications

Quantity 1 g
Formula Weight 151.23
Percent Purity 97%
Chemical Name or Material 3,4-Dihydro-2H-1,4-benzothiazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only