missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Difluorophenylacetic Acid 97.0+%, TCI America™
$55.92 - $167.76
Chemical Identifiers
| CAS | 658-93-5 |
|---|---|
| Molecular Formula | C8H6F2O2 |
| Molecular Weight (g/mol) | 172.131 |
| MDL Number | MFCD00010002 |
| InChI Key | YCAKYFIYUHHCKW-UHFFFAOYSA-N |
| Synonym | 3,4-difluorophenylacetic acid, 2-3,4-difluorophenyl acetic acid, 3,4-difluorophenyl acetic acid, benzeneacetic acid, 3,4-difluoro, 3,4-difluorobenzeneacetic acid, 3,4-difluorophenylaceticacid, pubchem4154, acmc-209nst, ksc355a2l, rarechem al bo 0250 |
| PubChem CID | 69571 |
| IUPAC Name | 2-(3,4-difluorophenyl)acetic acid |
| SMILES | C1=CC(=C(C=C1CC(=O)O)F)F |
Chemical Identifiers
| 658-93-5 | |
| 172.131 | |
| YCAKYFIYUHHCKW-UHFFFAOYSA-N | |
| 69571 | |
| C1=CC(=C(C=C1CC(=O)O)F)F |
| C8H6F2O2 | |
| MFCD00010002 | |
| 3,4-difluorophenylacetic acid, 2-3,4-difluorophenyl acetic acid, 3,4-difluorophenyl acetic acid, benzeneacetic acid, 3,4-difluoro, 3,4-difluorobenzeneacetic acid, 3,4-difluorophenylaceticacid, pubchem4154, acmc-209nst, ksc355a2l, rarechem al bo 0250 | |
| 2-(3,4-difluorophenyl)acetic acid |
Specifications
| 658-93-5 | |
| White-Yellow | |
| MFCD00010002 | |
| 3,4-difluorophenylacetic acid, 2-3,4-difluorophenyl acetic acid, 3,4-difluorophenyl acetic acid, benzeneacetic acid, 3,4-difluoro, 3,4-difluorobenzeneacetic acid, 3,4-difluorophenylaceticacid, pubchem4154, acmc-209nst, ksc355a2l, rarechem al bo 0250 | |
| C1=CC(=C(C=C1CC(=O)O)F)F | |
| 172.131 | |
| 172.13 | |
| Crystalline Powder |
| 50°C | |
| C8H6F2O2 | |
| 1 g | |
| YCAKYFIYUHHCKW-UHFFFAOYSA-N | |
| 2-(3,4-difluorophenyl)acetic acid | |
| 69571 | |
| ≥97.0% (GC,T) | |
| 3,4-Difluorophenylacetic Acid |
Safety and Handling
TSCA : No