missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Difluorophenylacetic Acid 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 50°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 172.13 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,4-Difluorophenylacetic Acid |
Safety and Handling
| TSCA | No |