Learn More
3,4-Difluorophenylacetic acid, 98%, Thermo Scientific Chemicals
$255.97 - $255.97
Chemical Identifiers
| CAS | 658-93-5 |
|---|---|
| Molecular Formula | C8H6F2O2 |
| Molecular Weight (g/mol) | 172.131 |
| MDL Number | MFCD00010002 |
| InChI Key | YCAKYFIYUHHCKW-UHFFFAOYSA-N |
| Synonym | 3,4-difluorophenylacetic acid, 2-3,4-difluorophenyl acetic acid, 3,4-difluorophenyl acetic acid, benzeneacetic acid, 3,4-difluoro, 3,4-difluorobenzeneacetic acid, 3,4-difluorophenylaceticacid, pubchem4154, acmc-209nst, ksc355a2l, rarechem al bo 0250 |
| PubChem CID | 69571 |
| IUPAC Name | 2-(3,4-difluorophenyl)acetic acid |
| SMILES | C1=CC(=C(C=C1CC(=O)O)F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1658406
|
Thermo Scientific Chemicals
A1658406 |
5 g |
Each for $255.97
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 658-93-5 | |
| 172.131 | |
| YCAKYFIYUHHCKW-UHFFFAOYSA-N | |
| 69571 | |
| C1=CC(=C(C=C1CC(=O)O)F)F |
| C8H6F2O2 | |
| MFCD00010002 | |
| 3,4-difluorophenylacetic acid, 2-3,4-difluorophenyl acetic acid, 3,4-difluorophenyl acetic acid, benzeneacetic acid, 3,4-difluoro, 3,4-difluorobenzeneacetic acid, 3,4-difluorophenylaceticacid, pubchem4154, acmc-209nst, ksc355a2l, rarechem al bo 0250 | |
| 2-(3,4-difluorophenyl)acetic acid |
Specifications
| 658-93-5 | |
| >110°C (230°F) | |
| MFCD00010002 | |
| 2577768 | |
| YCAKYFIYUHHCKW-UHFFFAOYSA-N | |
| 2-(3,4-difluorophenyl)acetic acid | |
| 69571 | |
| 98% |
| 48°C to 50°C | |
| C8H6F2O2 | |
| 5 g | |
| 3,4-difluorophenylacetic acid, 2-3,4-difluorophenyl acetic acid, 3,4-difluorophenyl acetic acid, benzeneacetic acid, 3,4-difluoro, 3,4-difluorobenzeneacetic acid, 3,4-difluorophenylaceticacid, pubchem4154, acmc-209nst, ksc355a2l, rarechem al bo 0250 | |
| C1=CC(=C(C=C1CC(=O)O)F)F | |
| 172.131 | |
| 172.13 | |
| 3,4-Difluorophenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-527-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only