missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAA1658406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1658406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1658406 Supplier Thermo Scientific Chemicals Supplier No. A1658406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 658-93-5 | Molecular Formula: C8H6F2O2 | Molecular Weight: 172.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 658-93-5
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00010002
InChI Key YCAKYFIYUHHCKW-UHFFFAOYSA-N
Synonym 3,4-difluorophenylacetic acid, 2-3,4-difluorophenyl acetic acid, 3,4-difluorophenyl acetic acid, benzeneacetic acid, 3,4-difluoro, 3,4-difluorobenzeneacetic acid, 3,4-difluorophenylaceticacid, pubchem4154, acmc-209nst, ksc355a2l, rarechem al bo 0250
PubChem CID 69571
IUPAC Name 2-(3,4-difluorophenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)F)F

Specifications

Melting Point 48°C to 50°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2577768
Formula Weight 172.13
Percent Purity 98%
Chemical Name or Material 3,4-Difluorophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-527-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only