missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorophenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. p-7128285
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44581G 1 g
D44585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44581G Supplier TCI America Supplier No. D44581G
Only null left

Chemical Identifiers

CAS 113028-75-4
Molecular Formula C7H3F2NS
Molecular Weight (g/mol) 171.16
MDL Number MFCD00175109
InChI Key SVZKYXSJICYUOH-UHFFFAOYSA-N
Synonym Isothiocyanic Acid 3,4-Difluorophenyl Ester
PubChem CID 2758331
IUPAC Name 1,2-difluoro-4-isothiocyanatobenzene
SMILES FC1=C(F)C=C(C=C1)N=C=S

Specifications

Color Yellow
Boiling Point 170°C
Quantity 1 g
UN Number 2810
Formula Weight 171.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Difluorophenyl Isothiocyanate