missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Difluorophenol 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| UN Number | 1325 |
| Formula Weight | 130.09 |
| Percent Purity | ≥98.0% (GC) |
| Chemical Name or Material | 3,4-Difluorophenol |
Safety and Handling
| TSCA | No |