missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorophenol 98.0+%, TCI America™

Catalog No. D24765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24765G 5 g
D247625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24765G Supplier TCI America Supplier No. D24765G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2713-33-9
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.09
MDL Number MFCD00010315
InChI Key BNPWVUJOPCGHIK-UHFFFAOYSA-N
Synonym phenol, 3,4-difluoro, 3,4-difluoro phenol, 3 4-difluorophenol, 3,4-difluoro-phenol, pubchem2315, acmc-1cmaj, 3,4-difluorophenol, ksc202q5j, bnpwvujopcghik-uhfffaoysa
PubChem CID 75927
IUPAC Name 3,4-difluorophenol
SMILES OC1=CC=C(F)C(F)=C1

Specifications

Quantity 5 g
UN Number 1325
Formula Weight 130.09
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3,4-Difluorophenol