missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzoyl chloride, 98%

Catalog No. AAL0818806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0818806 5 g
AAL0818814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0818806 Supplier Thermo Scientific Chemicals Supplier No. L0818806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 76903-88-3 | C7H3ClF2O | 176.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76903-88-3
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.55
MDL Number MFCD00009930
InChI Key RPQWXGVZELKOEU-UHFFFAOYSA-N
Synonym 3,4-difluorobenzoylchloride, benzoyl chloride, 3,4-difluoro, 3,4-difluoro benzoyl chloride, pubchem14796, acmc-209p6r, 3,4 difluorobenzoylchloride, 3,4-diflorobenzoyl chloride, 3,4-difluorobezoyl chloride, 3,4-diflourobenzoyl chloride, ksc494e2f
PubChem CID 2724519
IUPAC Name 3,4-difluorobenzoyl chloride
SMILES FC1=CC=C(C=C1F)C(Cl)=O

Specifications

Density 1.402
Boiling Point 185°C to 186°C
Flash Point 78°C (172°F)
Refractive Index 1.511
Quantity 5 g
UN Number UN3265
Beilstein 4177619
Sensitivity Moisture sensitive
Formula Weight 176.55
Percent Purity 98%
Chemical Name or Material 3,4-Difluorobenzoyl chloride
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 278-570-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only