missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzoyl Chloride 95.0+%, TCI America™

Catalog No. D28475G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28475G Supplier TCI America Supplier No. D28475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76903-88-3
Molecular Formula C7H3ClF2O
Molecular Weight (g/mol) 176.55
MDL Number MFCD00009930
InChI Key RPQWXGVZELKOEU-UHFFFAOYSA-N
Synonym 3,4-difluorobenzoylchloride, benzoyl chloride, 3,4-difluoro, 3,4-difluoro benzoyl chloride, pubchem14796, acmc-209p6r, 3,4 difluorobenzoylchloride, 3,4-diflorobenzoyl chloride, 3,4-difluorobezoyl chloride, 3,4-diflourobenzoyl chloride, ksc494e2f
PubChem CID 2724519
IUPAC Name 3,4-difluorobenzoyl chloride
SMILES FC1=CC=C(C=C1F)C(Cl)=O

Specifications

Color Yellow
Boiling Point 44°C
Quantity 5 g
UN Number 3265
Formula Weight 176.55
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Difluorobenzoyl Chloride