missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzotrifluoride 97.0+%, TCI America™

Catalog No. D33705G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D33705G Supplier TCI America Supplier No. D33705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32137-19-2
Molecular Formula C7H3F5
Molecular Weight (g/mol) 182.09
MDL Number MFCD00077510
InChI Key MVCGQTYWLZSKSB-UHFFFAOYSA-N
Synonym 3,4-difluorobenzotrifluoride, 1,2-difluoro-4-trifluoromethyl benzene, alpha,alpha,alpha,3,4-pentafluorotoluene, benzene, 1,2-difluoro-4-trifluoromethyl, pubchem1651, ksc497k4r, 1,2-difluoro-4-trifluoromethyl benzene #
PubChem CID 602797
IUPAC Name 1,2-difluoro-4-(trifluoromethyl)benzene
SMILES FC1=CC=C(C=C1F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 g
UN Number 1993
Formula Weight 182.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Difluorobenzotrifluoride