missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzophenone 98.0+%, TCI America™

Catalog No. p-7127798
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35765G 5 g
D357625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35765G Supplier TCI America Supplier No. D35765G
Only null left

Chemical Identifiers

CAS 85118-07-6
Molecular Formula C13H8F2O
Molecular Weight (g/mol) 218.203
MDL Number MFCD00009892
InChI Key ZJTYHSBOZAQQGF-UHFFFAOYSA-N
Synonym 3,4-difluorobenzophenone, 3,4-difluorophenyl phenyl methanone, methanone, 3,4-difluorophenyl phenyl, 3,4-difluoro-phenyl-phenyl-methanone, 3,4-difluorophenyl phenyl ketone, fr bf dvr, pubchem4220, acmc-209q4q, timtec-bb sbb016583
PubChem CID 569908
IUPAC Name (3,4-difluorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=C(C=C2)F)F

Specifications

Melting Point 55°C
Quantity 5 g
Formula Weight 218.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Difluorobenzophenone