missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Difluorobenzonitrile 98.0+%, TCI America™
$87.60 - $260.97
Chemical Identifiers
| CAS | 64248-62-0 |
|---|---|
| Molecular Formula | C7H3F2N |
| Molecular Weight (g/mol) | 139.11 |
| MDL Number | MFCD00011666 |
| InChI Key | BTBFCBQZFMQBNT-UHFFFAOYSA-N |
| Synonym | benzonitrile, 3,4-difluoro, 4-cyano-1,2-difluorobenzene, 3,4-difluoro benzonitrile, 3,4-difluorobenzenecarbonitrile, pubchem1562, acmc-209nkz, 3,4 difluorobenzonitrile, 3,4-difluoro-benzonitrile, ksc355i7l, 4-fluoro-3-fluorobenzonitrile |
| PubChem CID | 587203 |
| IUPAC Name | 3,4-difluorobenzonitrile |
| SMILES | FC1=CC=C(C=C1F)C#N |
Chemical Identifiers
| 64248-62-0 | |
| 139.11 | |
| BTBFCBQZFMQBNT-UHFFFAOYSA-N | |
| 587203 | |
| FC1=CC=C(C=C1F)C#N |
| C7H3F2N | |
| MFCD00011666 | |
| benzonitrile, 3,4-difluoro, 4-cyano-1,2-difluorobenzene, 3,4-difluoro benzonitrile, 3,4-difluorobenzenecarbonitrile, pubchem1562, acmc-209nkz, 3,4 difluorobenzonitrile, 3,4-difluoro-benzonitrile, ksc355i7l, 4-fluoro-3-fluorobenzonitrile | |
| 3,4-difluorobenzonitrile |
Specifications
| 64248-62-0 | |
| White-Yellow | |
| C7H3F2N | |
| 5 g | |
| benzonitrile, 3,4-difluoro, 4-cyano-1,2-difluorobenzene, 3,4-difluoro benzonitrile, 3,4-difluorobenzenecarbonitrile, pubchem1562, acmc-209nkz, 3,4 difluorobenzonitrile, 3,4-difluoro-benzonitrile, ksc355i7l, 4-fluoro-3-fluorobenzonitrile | |
| FC1=CC=C(C=C1F)C#N | |
| 139.11 | |
| 139.10 | |
| Crystalline Powder |
| 50°C | |
| 95°C | |
| MFCD00011666 | |
| 3439 | |
| BTBFCBQZFMQBNT-UHFFFAOYSA-N | |
| 3,4-difluorobenzonitrile | |
| 587203 | |
| ≥98.0% (GC) | |
| 3,4-Difluorobenzonitrile |
Safety and Handling
EINECSNumber : (3)-4305
TSCA : No