missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzonitrile, 98%

CAS: 64248-62-0 | C7H3F2N | 139.11 g/mol

$106.44 - $1150.97

Chemical Identifiers

CAS 64248-62-0
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.11
MDL Number MFCD00011666
InChI Key BTBFCBQZFMQBNT-UHFFFAOYSA-N
Synonym benzonitrile, 3,4-difluoro, 4-cyano-1,2-difluorobenzene, 3,4-difluoro benzonitrile, 3,4-difluorobenzenecarbonitrile, pubchem1562, acmc-209nkz, 3,4 difluorobenzonitrile, 3,4-difluoro-benzonitrile, ksc355i7l, 4-fluoro-3-fluorobenzonitrile
PubChem CID 587203
IUPAC Name 3,4-difluorobenzonitrile
SMILES FC1=CC=C(C=C1F)C#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1560006
View Documents
Thermo Scientific Chemicals
A1560006
5 g
Each for $106.44
Only null left
 
AAA1560014
View Documents
Thermo Scientific Chemicals
A1560014
25 g
Each for $324.82
Only null left
 
AAA1560022
View Documents
Thermo Scientific Chemicals
A1560022
100 g
Each for $1,150.97
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

64248-62-0
139.11
BTBFCBQZFMQBNT-UHFFFAOYSA-N
587203
FC1=CC=C(C=C1F)C#N
C7H3F2N
MFCD00011666
benzonitrile, 3,4-difluoro, 4-cyano-1,2-difluorobenzene, 3,4-difluoro benzonitrile, 3,4-difluorobenzenecarbonitrile, pubchem1562, acmc-209nkz, 3,4 difluorobenzonitrile, 3,4-difluoro-benzonitrile, ksc355i7l, 4-fluoro-3-fluorobenzonitrile
3,4-difluorobenzonitrile

Specifications

64248-62-0
180°C
C7H3F2N
5 g
2082224
BTBFCBQZFMQBNT-UHFFFAOYSA-N
3,4-difluorobenzonitrile
587203
98%
48°C to 52°C
69°C (156°F)
MFCD00011666
UN3439
benzonitrile, 3,4-difluoro, 4-cyano-1,2-difluorobenzene, 3,4-difluoro benzonitrile, 3,4-difluorobenzenecarbonitrile, pubchem1562, acmc-209nkz, 3,4 difluorobenzonitrile, 3,4-difluoro-benzonitrile, ksc355i7l, 4-fluoro-3-fluorobenzonitrile
FC1=CC=C(C=C1F)C#N
139.11
139.11
3,4-Difluorobenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 264-751-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only