missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Difluorobenzoic Acid 98.0+%, TCI America™
$136.08 - $432.46
Chemical Identifiers
| CAS | 455-86-7 |
|---|---|
| Molecular Formula | C7H4F2O2 |
| Molecular Weight (g/mol) | 158.104 |
| MDL Number | MFCD00011672 |
| InChI Key | FPENCTDAQQQKNY-UHFFFAOYSA-N |
| Synonym | benzoic acid, 3,4-difluoro, 3,4-fifluorobenzoic acid, 3,4-difluoro-benzoic acid, 3,4-dichloropnenol, pubchem1337, acmc-209k3h, 3,4-difluoro benzoic acid, ksc237g8t, rarechem al bo 0269, timtec-bb sbb006721 |
| PubChem CID | 99166 |
| IUPAC Name | 3,4-difluorobenzoic acid |
| SMILES | C1=CC(=C(C=C1C(=O)O)F)F |
Chemical Identifiers
| 455-86-7 | |
| 158.104 | |
| FPENCTDAQQQKNY-UHFFFAOYSA-N | |
| 99166 | |
| C1=CC(=C(C=C1C(=O)O)F)F |
| C7H4F2O2 | |
| MFCD00011672 | |
| benzoic acid, 3,4-difluoro, 3,4-fifluorobenzoic acid, 3,4-difluoro-benzoic acid, 3,4-dichloropnenol, pubchem1337, acmc-209k3h, 3,4-difluoro benzoic acid, ksc237g8t, rarechem al bo 0269, timtec-bb sbb006721 | |
| 3,4-difluorobenzoic acid |
Specifications
| 455-86-7 | |
| White | |
| MFCD00011672 | |
| benzoic acid, 3,4-difluoro, 3,4-fifluorobenzoic acid, 3,4-difluoro-benzoic acid, 3,4-dichloropnenol, pubchem1337, acmc-209k3h, 3,4-difluoro benzoic acid, ksc237g8t, rarechem al bo 0269, timtec-bb sbb006721 | |
| C1=CC(=C(C=C1C(=O)O)F)F | |
| 158.104 | |
| 158.10 | |
| Crystalline Powder |
| 123°C | |
| C7H4F2O2 | |
| 5 g | |
| FPENCTDAQQQKNY-UHFFFAOYSA-N | |
| 3,4-difluorobenzoic acid | |
| 99166 | |
| ≥98.0% (GC,T) | |
| 3,4-Difluorobenzoic Acid |
Safety and Handling
TSCA : No