missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzoic acid, 98%

CAS: 455-86-7 | C7H4F2O2 | 158.104 g/mol

$36.90 - $370.71

Chemical Identifiers

CAS 455-86-7
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD00011672
InChI Key FPENCTDAQQQKNY-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-difluoro, 3,4-fifluorobenzoic acid, 3,4-difluoro-benzoic acid, 3,4-dichloropnenol, pubchem1337, acmc-209k3h, 3,4-difluoro benzoic acid, ksc237g8t, rarechem al bo 0269, timtec-bb sbb006721
PubChem CID 99166
IUPAC Name 3,4-difluorobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1587603
View Documents
Thermo Scientific Chemicals
A1587603
1 g
Each for $36.90
Only null left
 
AAA1587606
View Documents
Thermo Scientific Chemicals
A1587606
5 g
Each for $111.74
Only null left
 
AAA1587614
View Documents
Thermo Scientific Chemicals
A1587614
25 g
Each for $370.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

455-86-7
158.104
FPENCTDAQQQKNY-UHFFFAOYSA-N
99166
C1=CC(=C(C=C1C(=O)O)F)F
C7H4F2O2
MFCD00011672
benzoic acid, 3,4-difluoro, 3,4-fifluorobenzoic acid, 3,4-difluoro-benzoic acid, 3,4-dichloropnenol, pubchem1337, acmc-209k3h, 3,4-difluoro benzoic acid, ksc237g8t, rarechem al bo 0269, timtec-bb sbb006721
3,4-difluorobenzoic acid

Specifications

455-86-7
Odorless
MFCD00011672
2085848
FPENCTDAQQQKNY-UHFFFAOYSA-N
3,4-difluorobenzoic acid
99166
98%
121°C to 124°C
C7H4F2O2
1 g
benzoic acid, 3,4-difluoro, 3,4-fifluorobenzoic acid, 3,4-difluoro-benzoic acid, 3,4-dichloropnenol, pubchem1337, acmc-209k3h, 3,4-difluoro benzoic acid, ksc237g8t, rarechem al bo 0269, timtec-bb sbb006721
C1=CC(=C(C=C1C(=O)O)F)F
158.104
158.11
3,4-Difluorobenzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-249-6

TSCA : Yes

SDS
Documents

Documents

RUO – Research Use Only