missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Difluorobenzenesulfonamide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 88°C |
| Color | White |
| Quantity | 5 g |
| Synonym | 3,4-difluorobenzenesulphonamide, 3,4-difluorobenzene-1-sulfonamide, 3,4-difluoro-benzenesulfonamide, benzenesulfonamide, 3,4-difluoro, acmc-20ahuj, pubchem11788, benzenesulfonamide,3,4-difluoro, 3,4-difluorobenzenesulfonamide, 3,4-difluoro benzene sulfonamide, benzenesulfonamide, 3,4-difluoro-9ci |
| Formula Weight | 193.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,4-Difluorobenzenesulfonamide |
Safety and Handling
| TSCA | No |