missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorothiophenol, 97%

Catalog No. AAL1203106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1203106 5 g
AAL1203103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1203106 Supplier Thermo Scientific Chemicals Supplier No. L1203106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5858-17-3 | C6H4Cl2S | 179.058 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5858-17-3
Molecular Formula C6H4Cl2S
Molecular Weight (g/mol) 179.058
MDL Number MFCD00004840
InChI Key HNJZDPKMMZXSKT-UHFFFAOYSA-N
Synonym 3,4-dichlorothiophenol, 3,4-dichloro thiophenol, benzenethiol, 3,4-dichloro, 3,4-dichlorobenzene-1-thiol, pubchem6803, 3,4dichloro-thiophenol, acmc-1aset, 3,4-dichloro-thiophenol, 3 ,4-dichlorobenzenethiol, 3,4-dichlorophenylmercaptan
PubChem CID 79964
IUPAC Name 3,4-dichlorobenzenethiol
SMILES C1=CC(=C(C=C1S)Cl)Cl

Specifications

Density 1.407
Boiling Point 119°C to 120°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.623
Quantity 5 g
Beilstein 2042869
Sensitivity Air Sensitive
Formula Weight 179.07
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorothiophenol
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 227-485-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only