missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenylhydrazine hydrochloride, 98+%

Catalog No. AAA1390609
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1390609 10 g
AAA1390618 50 g
AAA1390630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1390609 Supplier Thermo Scientific Chemicals Supplier No. A1390609

CAS: 19763-90-7 | C6H7Cl3N2 | 213.486 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19763-90-7
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
MDL Number MFCD00012937
InChI Key WIKZRYSGQPZTMM-UHFFFAOYSA-N
Synonym 3,4-dichlorophenylhydrazine hydrochloride, 3,4-dichlorophenyl hydrazine hydrochloride, 1-3,4-dichlorophenyl hydrazine hydrochloride, 3,4-dichlorophenyl hydrazine monohydrochloride, hydrazine, 3,4-dichlorophenyl-, monohydrochloride, 3,4-dichlorophenylhydrazinehydrochloride, pubchem7530, acmc-209f1i, ksc491e2j
PubChem CID 3084323
IUPAC Name (3,4-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=C(C=C1NN)Cl)Cl.Cl

Specifications

Melting Point ∼229°C (decomposition)
Quantity 10 g
Beilstein 3703934
Formula Weight 213.5
Percent Purity ≥98%
Chemical Name or Material 3,4-Dichlorophenylhydrazine hydrochloride
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 243-278-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only