missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloronitrobenzene 97.0+%, TCI America™

Catalog No. D0388500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0388500G 500 g
D038825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0388500G Supplier TCI America Supplier No. D0388500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-54-7
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00007207
InChI Key NTBYINQTYWZXLH-UHFFFAOYSA-N
Synonym 3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, dcnb, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, unii-ijt74ji7fs, ccris 3097, 1,2-dichloro-4-nitro-benzene, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech
PubChem CID 7443
IUPAC Name 1,2-dichloro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 41°C
Boiling Point 255°C
Quantity 500 g
UN Number 1578
Formula Weight 192.00
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dichloronitrobenzene
EINECSNumber (3)-0455
RTECSNumber CZ5250000
TSCA Yes