Learn More
3,4-Dichloronitrobenzene, 95%, Thermo Scientific™
$90.59 - $90.59
Chemical Identifiers
| CAS | 99-54-7 |
|---|---|
| Molecular Formula | C6H3Cl2NO2 |
| Molecular Weight (g/mol) | 192.00 |
| MDL Number | MFCD00007207 |
| InChI Key | NTBYINQTYWZXLH-UHFFFAOYSA-N |
| Synonym | 3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, dcnb, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, unii-ijt74ji7fs, ccris 3097, 1,2-dichloro-4-nitro-benzene, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech |
| PubChem CID | 7443 |
| IUPAC Name | 1,2-dichloro-4-nitrobenzene |
| SMILES | [O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC113532500
|
Thermo Scientific Chemicals
113532500 |
250 g | Glass bottle |
Each for $90.59
|
|
||||
Chemical Identifiers
| 99-54-7 | |
| 192.00 | |
| NTBYINQTYWZXLH-UHFFFAOYSA-N | |
| 7443 | |
| [O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1 |
| C6H3Cl2NO2 | |
| MFCD00007207 | |
| 3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, dcnb, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, unii-ijt74ji7fs, ccris 3097, 1,2-dichloro-4-nitro-benzene, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech | |
| 1,2-dichloro-4-nitrobenzene |
Specifications
| 99-54-7 | |
| 100.0 | |
| Yellow to Brown | |
| 95% | |
| C6H3Cl2NO2 | |
| MFCD00007207 | |
| 05, 246 | |
| NTBYINQTYWZXLH-UHFFFAOYSA-N | |
| 1,2-dichloro-4-nitrobenzene | |
| 7443 | |
| ≥94% | |
| 3, 4-Dichloronitrobenzene, 95% |
| 94.0 | |
| 39°C to 45°C | |
| Authentic | |
| Glass bottle | |
| Cl2C6H3NO2 | |
| 250 g | |
| 3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, dcnb, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, unii-ijt74ji7fs, ccris 3097, 1,2-dichloro-4-nitro-benzene, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech | |
| [O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1 | |
| 192.00 | |
| 192 | |
| Crystalline Mass |
Safety and Handling
GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat, drink or smoke when using this product.
Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 202-764-2
RTECSNumber : CZ5250000
TSCA : TSCA
RUO – Research Use Only