missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloronitrobenzene, 95%, Thermo Scientific™

Catalog No. AC113532500
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113532500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC113532500 Supplier Thermo Scientific Chemicals Supplier No. 113532500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-54-7
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00007207
InChI Key NTBYINQTYWZXLH-UHFFFAOYSA-N
Synonym 3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, dcnb, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, unii-ijt74ji7fs, ccris 3097, 1,2-dichloro-4-nitro-benzene, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech
PubChem CID 7443
IUPAC Name 1,2-dichloro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 39°C to 45°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula Cl2C6H3NO2
Quantity 250 g
Beilstein 05, 246
Formula Weight 192
Percent Purity ≥94%
Physical Form Crystalline Mass
Chemical Name or Material 3, 4-Dichloronitrobenzene, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat, drink or smoke when using this product.
Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-764-2
RTECSNumber CZ5250000
TSCA TSCA

RUO – Research Use Only