missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzylamine, 98%

Catalog No. AC113240050
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113240050 5 mL Glass bottle
AC113240250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC113240050 Supplier Thermo Scientific Chemicals Supplier No. 113240050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 102-49-8 | C7H7Cl2N | 176.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-49-8
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.05
MDL Number MFCD00008114
InChI Key IXHNFOOSLAWRBQ-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzylamine, 3,4-dichlorophenyl methanamine, benzenemethanamine, 3,4-dichloro, 3,4-dichloro-benzylamine, benzylamine, 3,4-dichloro, 3,4-dichlorophenyl methylamine, 1-3,4-dichlorophenyl methanamine, 3,4-dichlorophenyl methyl amine, 3,4-dichlorobezylamine
PubChem CID 1608
IUPAC Name (3,4-dichlorophenyl)methanamine
SMILES C1=CC(=C(C=C1CN)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3200g/mL
Boiling Point 80°C to 82°C (0.6 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5775 to 1.5795
Linear Formula Cl2C6H3CH2NH2
Quantity 5 mL
Specific Gravity 1.32
Formula Weight 176.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3, 4-Dichlorobenzylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-035-1
TSCA TSCA

RUO – Research Use Only