missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzylamine, 96%

Catalog No. AAL0489906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0489906 5 g
AAL0489914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0489906 Supplier Thermo Scientific Chemicals Supplier No. L0489906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 102-49-8 | C7H7Cl2N | 176.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-49-8
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00008114
InChI Key IXHNFOOSLAWRBQ-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzylamine, 3,4-dichlorophenyl methanamine, benzenemethanamine, 3,4-dichloro, 3,4-dichloro-benzylamine, benzylamine, 3,4-dichloro, 3,4-dichlorophenyl methylamine, 1-3,4-dichlorophenyl methanamine, 3,4-dichlorophenyl methyl amine, 3,4-dichlorobezylamine
PubChem CID 1608
IUPAC Name (3,4-dichlorophenyl)methanamine
SMILES C1=CC(=C(C=C1CN)Cl)Cl

Specifications

Density 1.322
Boiling Point 80°C to 82°C (0.6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.578
Quantity 5 g
UN Number UN2735
Beilstein 508818
Sensitivity Air Sensitive
Formula Weight 176.05
Percent Purity 96%
Chemical Name or Material 3,4-Dichlorobenzylamine
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 203-035-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only