missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl isocyanate, 98%

Catalog No. p-3778193
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC319220010 1 g Glass bottle
AC319220100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC319220010 Supplier Thermo Scientific Chemicals Supplier No. 319220010
Only null left

CAS: 19752-09-1 | C8H5Cl2NO | 202.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19752-09-1
Molecular Formula C8H5Cl2NO
Molecular Weight (g/mol) 202.04
MDL Number MFCD00673063
InChI Key WLNAHQZFPSSVTP-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl isocyanate, 3,4-dichlorobenzylisocyanate, 1,2-dichloro-4-isocyanatomethyl benzene, benzene, 1,2-dichloro-4-isocyanatomethyl, 1,2-dichloro-4-isocyanatomethyl-benzene, 3,4-dichlorophenyl methanisocyanate, zlchem 123, acmc-20anfi, pubchem2096, 3,4-dichloro-benzyl isocyanate
PubChem CID 2733368
IUPAC Name 1,2-dichloro-4-(isocyanatomethyl)benzene
SMILES C1=CC(=C(C=C1CN=C=O)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.3750g/mL
Boiling Point 282°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5656 to 1.5676
Quantity 1 g
Specific Gravity 1.375
Formula Weight 202.04
Percent Purity 98%
Physical Form Slightly Viscous Liquid
Chemical Name or Material 3, 4-Dichlorobenzyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only