accessibility menu, dialog, popup

UserName

ClNCOCl

CAS RN 19752-09-1

IUPAC Name: 1,2-dichloro-4-(isocyanatomethyl)benzene
Synonyms: 1,2-dichloro-4-(isocyanatomethyl)benzene
Molecular Weight (g/mol): 202.03
Molecular Formula: C8H5Cl2NO
InChi Key: WLNAHQZFPSSVTP-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CN=C=O)C=C1Cl
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results