missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl chloride, 98%

Catalog No. AAA1767214
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1767214 25 g
AAA1767218 50 g
AAA1767222 100 g
AAA1767230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1767214 Supplier Thermo Scientific Chemicals Supplier No. A1767214
Only null left

CAS: 102-47-6 | C7H5Cl3 | 195.467 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-47-6
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.467
MDL Number MFCD00000906
InChI Key YZIFVWOCPGPNHB-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl chloride, 1,2-dichloro-4-chloromethyl benzene, alpha,3,4-trichlorotoluene, benzene, 1,2-dichloro-4-chloromethyl, unii-451pb8smie, 1,2-dichloro-4-chloromethyl-benzene, 3,4-dichlorobenzylchloride, 451pb8smie, 3,4-dichlorophenylmethyl chloride, toluene, .alpha.,3,4-trichloro
PubChem CID 7609
IUPAC Name 1,2-dichloro-4-(chloromethyl)benzene
SMILES C1=CC(=C(C=C1CCl)Cl)Cl

Specifications

Melting Point -3°C
Density 1.416
Boiling Point 99°C to 101°C (2 mmHg)
Flash Point 155°C (311°F)
Odor Pungent
Refractive Index 1.5766
Quantity 25 g
UN Number UN3265
Beilstein 386644
Formula Weight 195.48
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorobenzyl chloride
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 203-033-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only