missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl Bromide 98.0+%, TCI America™

Catalog No. D37005G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37005G 5 g
D370025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37005G Supplier TCI America Supplier No. D37005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18880-04-1
Molecular Formula C7H5BrCl2
Molecular Weight (g/mol) 239.921
MDL Number MFCD00041363
InChI Key XLWSBDFQAJXCQX-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl bromide, 4-bromomethyl-1,2-dichlorobenzene, benzene, 4-bromomethyl-1,2-dichloro, 3,4-dichlorobenzylbromide, alpha-bromo-3,4-dichlorotoluene, acmc-209erq, 3,4 dichlorobenzyl bromide, 3,4-dichloro-benzyl bromide, ksc493a6b, timtec-bb sbb005763
PubChem CID 639726
IUPAC Name 4-(bromomethyl)-1,2-dichlorobenzene
SMILES C1=CC(=C(C=C1CBr)Cl)Cl

Specifications

Color Yellow
Boiling Point 86°C
Quantity 5 g
UN Number 3265
Formula Weight 239.92
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dichlorobenzyl Bromide