missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl bromide, 98%

Catalog No. AAL0525406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0525406 5 g
AAL0525414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0525406 Supplier Thermo Scientific Chemicals Supplier No. L0525406

CAS: 18880-04-1 | C7H5BrCl2 | 239.921 g/mol

3,4-Dichlorobenzyl bromide is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dichlorobenzyl bromide is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases.

Chemical Identifiers

CAS 18880-04-1
Molecular Formula C7H5BrCl2
Molecular Weight (g/mol) 239.921
MDL Number MFCD00041363
InChI Key XLWSBDFQAJXCQX-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl bromide, 4-bromomethyl-1,2-dichlorobenzene, benzene, 4-bromomethyl-1,2-dichloro, 3,4-dichlorobenzylbromide, alpha-bromo-3,4-dichlorotoluene, acmc-209erq, 3,4 dichlorobenzyl bromide, 3,4-dichloro-benzyl bromide, ksc493a6b, timtec-bb sbb005763
PubChem CID 639726
IUPAC Name 4-(bromomethyl)-1,2-dichlorobenzene
SMILES C1=CC(=C(C=C1CBr)Cl)Cl

Specifications

Density 1.65
Boiling Point 116°C to 119°C (1 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.6065
Quantity 5 g
UN Number UN3265
Beilstein 742579
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 239.93
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorobenzyl bromide
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only