missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzotrifluoride 98.0+%, TCI America™

Catalog No. D1865500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1865500G 500 g
D186525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1865500G Supplier TCI America Supplier No. D1865500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 328-84-7
Molecular Formula C7H3Cl2F3
Molecular Weight (g/mol) 214.996
MDL Number MFCD00000555
InChI Key XILPLWOGHPSJBK-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzotrifluoride, 1,2-dichloro-4-trifluoromethyl benzene, benzene, 1,2-dichloro-4-trifluoromethyl, 3,4-dichloro-alpha,alpha,alpha-trifluorotoluene, 3,4-dichlorotrifluoromethylbenzene, unii-7g60xcu4gg, 3,4-dichlorophenyltrifluoromethane, 7g60xcu4gg, 3,4-dichloro-1-trifluoromethyl benzene, 3,4-dcbte
PubChem CID 9481
IUPAC Name 1,2-dichloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)Cl)Cl

Specifications

Color Colorless
Boiling Point 173°C
Quantity 500 g
UN Number 1760
Formula Weight 215.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dichlorobenzotrifluoride
EINECSNumber (3)-3553
RTECSNumber CZ5527510
TSCA Yes