missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dichlorobenzenesulfonyl Chloride 98.0+%, TCI America™
Supplier: TCI America D299025G
Specifications
| 3,4-Dichlorobenzenesulfonyl Chloride | |
| White | |
| C6H3Cl3O2S | |
| 25 g | |
| 3,4-dichlorobenzene-1-sulfonyl chloride, 3,4-dichlorophenylsulfonyl chloride, 3,4-dichlorobenzenesulphonyl chloride, benzenesulfonyl chloride, 3,4-dichloro, 3,4-dichlorobenzenesulfonylchloride, 3,4-dichloro-benzenesulfonyl chloride, 3,4-dichlorobenzene-sulfonyl chloride, 3,4-dichloro benzene sulfonyl chloride, 3,4-dichlorophenyl chlorosulfone, pubchem5118 | |
| ClC1=CC=C(C=C1Cl)S(Cl)(=O)=O | |
| 245.50 | |
| 245.50 | |
| Wax |
| 98-31-7 | |
| 253°C | |
| MFCD00041255 | |
| 3261 | |
| NYIBPWGZGSXURD-UHFFFAOYSA-N | |
| 3,4-dichlorobenzene-1-sulfonyl chloride | |
| 7383 | |
| ≥98.0% (T) |
Chemical Identifiers
| 98-31-7 | |
| 245.50 | |
| NYIBPWGZGSXURD-UHFFFAOYSA-N | |
| 7383 | |
| ClC1=CC=C(C=C1Cl)S(Cl)(=O)=O |
| C6H3Cl3O2S | |
| MFCD00041255 | |
| 3,4-dichlorobenzene-1-sulfonyl chloride, 3,4-dichlorophenylsulfonyl chloride, 3,4-dichlorobenzenesulphonyl chloride, benzenesulfonyl chloride, 3,4-dichloro, 3,4-dichlorobenzenesulfonylchloride, 3,4-dichloro-benzenesulfonyl chloride, 3,4-dichlorobenzene-sulfonyl chloride, 3,4-dichloro benzene sulfonyl chloride, 3,4-dichlorophenyl chlorosulfone, pubchem5118 | |
| 3,4-dichlorobenzene-1-sulfonyl chloride |
Safety and Handling
TSCA : Yes