missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7127595
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29905G 5 g
D299025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29905G Supplier TCI America Supplier No. D29905G

Chemical Identifiers

CAS 98-31-7
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.50
MDL Number MFCD00041255
InChI Key NYIBPWGZGSXURD-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzene-1-sulfonyl chloride, 3,4-dichlorophenylsulfonyl chloride, 3,4-dichlorobenzenesulphonyl chloride, benzenesulfonyl chloride, 3,4-dichloro, 3,4-dichlorobenzenesulfonylchloride, 3,4-dichloro-benzenesulfonyl chloride, 3,4-dichlorobenzene-sulfonyl chloride, 3,4-dichloro benzene sulfonyl chloride, 3,4-dichlorophenyl chlorosulfone, pubchem5118
PubChem CID 7383
IUPAC Name 3,4-dichlorobenzene-1-sulfonyl chloride
SMILES ClC1=CC=C(C=C1Cl)S(Cl)(=O)=O

Specifications

Color White
Boiling Point 253°C
Quantity 5 g
UN Number 3261
Formula Weight 245.50
Percent Purity ≥98.0% (T)
Physical Form Wax
Chemical Name or Material 3,4-Dichlorobenzenesulfonyl Chloride