Learn More
3,4-Dichlorobenzeneboronic acid, 97%
CAS: 151169-75-4 | C6H5BCl2O2 | 190.81 g/mol
$28.25 - $317.75
Chemical Identifiers
| CAS | 151169-75-4 |
|---|---|
| Molecular Formula | C6H5BCl2O2 |
| Molecular Weight (g/mol) | 190.81 |
| MDL Number | MFCD01074646 |
| InChI Key | JKIGHOARKAIPJI-UHFFFAOYSA-N |
| Synonym | 3,4-dichlorophenyl boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenylboronicacid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenylbornic acid, chembl20776, 3,4-dichloro benzene boronic acid, boronic acid, 3,4-dichlorophenyl, pubchem1812 |
| PubChem CID | 2734330 |
| IUPAC Name | (3,4-dichlorophenyl)boronic acid |
| SMILES | OB(O)C1=CC=C(Cl)C(Cl)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2429203
|
Thermo Scientific Chemicals
B2429203 |
1 g |
Each for $28.25
|
|
|||||
|
AAB2429206
|
Thermo Scientific Chemicals
B2429206 |
5 g |
Each for $94.81
|
|
|||||
|
AAB2429214
|
Thermo Scientific Chemicals
B2429214 |
25 g |
Each for $317.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 151169-75-4 | |
| 190.81 | |
| JKIGHOARKAIPJI-UHFFFAOYSA-N | |
| 2734330 | |
| OB(O)C1=CC=C(Cl)C(Cl)=C1 |
| C6H5BCl2O2 | |
| MFCD01074646 | |
| 3,4-dichlorophenyl boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenylboronicacid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenylbornic acid, chembl20776, 3,4-dichloro benzene boronic acid, boronic acid, 3,4-dichlorophenyl, pubchem1812 | |
| (3,4-dichlorophenyl)boronic acid |
Specifications
| 151169-75-4 | |
| C6H5BCl2O2 | |
| 1 g | |
| 3,4-dichlorophenyl boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenylboronicacid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenylbornic acid, chembl20776, 3,4-dichloro benzene boronic acid, boronic acid, 3,4-dichlorophenyl, pubchem1812 | |
| OB(O)C1=CC=C(Cl)C(Cl)=C1 | |
| 190.81 | |
| 190.82 | |
| 3,4-Dichlorobenzeneboronic acid |
| 270°C to 274°C | |
| MFCD01074646 | |
| 7369790 | |
| JKIGHOARKAIPJI-UHFFFAOYSA-N | |
| (3,4-dichlorophenyl)boronic acid | |
| 2734330 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only