accessibility menu, dialog, popup

UserName

HOBOHClCl

CAS RN 151169-75-4

IUPAC Name: (3,4-dichlorophenyl)boronic acid
Synonyms: 3,4-dichlorophenylboronic acid
Molecular Weight (g/mol): 190.81
Molecular Formula: C6H5BCl2O2
InChi Key: JKIGHOARKAIPJI-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(Cl)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)