missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzaldehyde 98.0+%, TCI America™

Catalog No. p-7126152
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D033225G 25 g
D0332250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D033225G Supplier TCI America Supplier No. D033225G

Chemical Identifiers

CAS 6287-38-3
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00003351
InChI Key ZWUSBSHBFFPRNE-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-dichloro, unii-i6z632wsj5, ccris 6015, pubchem8230, 3,4-dichlorobenzaldehye, 3,4-dichlorobenzaldhyde, 3,4 dichlorobenzaldehyde, 3,4-dichlorobenzaidehyde, 4,3-dichlorobenzaldehyde, dsstox_cid_4971
PubChem CID 22710
IUPAC Name 3,4-dichlorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)Cl)Cl

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 248°C
Quantity 25 g
UN Number 1759
Formula Weight 175.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dichlorobenzaldehyde