accessibility menu, dialog, popup

UserName

CAS RN 6287-38-3

IUPAC Name: 3,4-dichlorobenzaldehyde
Synonyms: 3,4-dichlorobenzaldehyde
Molecular Weight (g/mol): 175.01
Molecular Formula: C7H4Cl2O
InChi Key: ZWUSBSHBFFPRNE-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=O)C=C1Cl
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)