missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dichloroaniline 98.0+%, TCI America™
Supplier: TCI America D0324500G
Specifications
| 3,4-Dichloroaniline | |
| 72°C | |
| 145°C | |
| MFCD00007768 | |
| 3442 | |
| SDYWXFYBZPNOFX-UHFFFAOYSA-N | |
| 3,4-dichloroaniline | |
| 7257 | |
| 162.01 | |
| Crystalline Powder |
| 95-76-1 | |
| White-Yellow | |
| C6H5Cl2N | |
| 500 g | |
| 3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline | |
| C1=CC(=C(C=C1N)Cl)Cl | |
| 162.013 | |
| CHEBI:16767 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 95-76-1 | |
| 162.013 | |
| SDYWXFYBZPNOFX-UHFFFAOYSA-N | |
| 7257 | |
| 3,4-dichloroaniline |
| C6H5Cl2N | |
| MFCD00007768 | |
| 3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline | |
| CHEBI:16767 | |
| C1=CC(=C(C=C1N)Cl)Cl |
Safety and Handling
EINECSNumber : (3)-0261
RTECSNumber : BX2625000
TSCA : Yes