missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloroaniline 98.0+%, TCI America™

Catalog No. p-7126145
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D032425G 25 g
D0324500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D032425G Supplier TCI America Supplier No. D032425G
Only null left

Chemical Identifiers

CAS 95-76-1
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00007768
InChI Key SDYWXFYBZPNOFX-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline
PubChem CID 7257
ChEBI CHEBI:16767
IUPAC Name 3,4-dichloroaniline
SMILES C1=CC(=C(C=C1N)Cl)Cl

Specifications

Melting Point 72°C
Color White-Yellow
Boiling Point 145°C
Quantity 25 g
UN Number 3442
Formula Weight 162.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dichloroaniline
EINECSNumber (3)-0261
RTECSNumber BX2625000
TSCA Yes