Learn More
3,4-Dichloroaniline, 98%
CAS: 95-76-1 | C6H5Cl2N | 162.013 g/mol
Supplier: Thermo Scientific Chemicals A1519930
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 3,4-Dichloroaniline | |
| 70°C to 73°C | |
| 135°C (275°F) | |
| MFCD00007768 | |
| UN3442 | |
| 14,3054 | |
| SDYWXFYBZPNOFX-UHFFFAOYSA-N | |
| 3,4-dichloroaniline | |
| 7257 | |
| 162.02 |
| 95-76-1 | |
| 272°C | |
| C6H5Cl2N | |
| 250 g | |
| 636837 | |
| 3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline | |
| C1=CC(=C(C=C1N)Cl)Cl | |
| 162.013 | |
| CHEBI:16767 | |
| 98% |
Chemical Identifiers
| 95-76-1 | |
| 162.013 | |
| SDYWXFYBZPNOFX-UHFFFAOYSA-N | |
| 7257 | |
| 3,4-dichloroaniline |
| C6H5Cl2N | |
| MFCD00007768 | |
| 3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline | |
| CHEBI:16767 | |
| C1=CC(=C(C=C1N)Cl)Cl |
Safety and Handling
GHS H Statement
H310-H330-H318-H302-H317
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H317-H318-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
EINECSNumber : 202-448-4
RTECSNumber : BX2625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only