missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloroaniline, 98%

Catalog No. p-7022425
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
2500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1519918 50 g
AAA1519930 250 g
AAA1519936 500 g
AAA151990E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1519918 Supplier Thermo Scientific Chemicals Supplier No. A1519918
Only null left

CAS: 95-76-1 | C6H5Cl2N | 162.013 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-76-1
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00007768
InChI Key SDYWXFYBZPNOFX-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline
PubChem CID 7257
ChEBI CHEBI:16767
IUPAC Name 3,4-dichloroaniline
SMILES C1=CC(=C(C=C1N)Cl)Cl

Specifications

Melting Point 70°C to 73°C
Boiling Point 272°C
Flash Point 135°C (275°F)
Quantity 50 g
UN Number UN3442
Beilstein 636837
Merck Index 14,3054
Formula Weight 162.02
Percent Purity 98%
Chemical Name or Material 3,4-Dichloroaniline
Hazard Category H301+H311+H331-H317-H318-H500
Hazard Statement GHS H Statement
H310-H330-H318-H302-H317
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
EINECSNumber 202-448-4
RTECSNumber BX2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only