missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dichloroacetophenone 98.0+%, TCI America™

Catalog No. D031125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D031125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D031125G Supplier TCI America Supplier No. D031125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2642-63-9
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00000553
InChI Key WBPAOUHWPONFEQ-UHFFFAOYSA-N
Synonym 3',4'-dichloroacetophenone, 1-3,4-dichlorophenyl ethanone, 3,4-dichloroacetophenone, 1-3,4-dichlorophenyl ethan-1-one, ethanone, 1-3,4-dichlorophenyl, acetophenone, 3',4'-dichloro, 1-acetyl-3,4-dichlorobenzene, 3,4 dichloro acetophenone, pubchem3383, 3,4-dicloroacetophenone
PubChem CID 75841
IUPAC Name 1-(3,4-dichlorophenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 72°C
Boiling Point 121°C
Quantity 25 g
Formula Weight 189.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,4′-Dichloroacetophenone