accessibility menu, dialog, popup

UserName

CAS RN 2642-63-9

IUPAC Name: 1-(3,4-dichlorophenyl)ethan-1-one
Synonyms: 1-(3,4-dichlorophenyl)ethanone
Molecular Weight (g/mol): 189.04
Molecular Formula: C8H6Cl2O
InChi Key: WBPAOUHWPONFEQ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(Cl)C(Cl)=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)