missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dichloroacetophenone, 98+%

Catalog No. AAA1356814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1356814 25 g
AAA1356822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1356814 Supplier Thermo Scientific Chemicals Supplier No. A1356814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2642-63-9 | C8H6Cl2O | 189.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2642-63-9
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00000553
InChI Key WBPAOUHWPONFEQ-UHFFFAOYSA-N
Synonym 3',4'-dichloroacetophenone, 1-3,4-dichlorophenyl ethanone, 3,4-dichloroacetophenone, 1-3,4-dichlorophenyl ethan-1-one, ethanone, 1-3,4-dichlorophenyl, acetophenone, 3',4'-dichloro, 1-acetyl-3,4-dichlorobenzene, 3,4 dichloro acetophenone, pubchem3383, 3,4-dicloroacetophenone
PubChem CID 75841
IUPAC Name 1-(3,4-dichlorophenyl)ethanone
SMILES CC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 72°C to 76°C
Boiling Point 134°C to 135 mmHg) (12 mmHg)
Quantity 25 g
Beilstein 742534
Formula Weight 189.04
Percent Purity ≥98%
Chemical Name or Material 3',4'-Dichloroacetophenone
Hazard Category H302
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 220-146-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only