missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dibromotoluene 98.0+%, TCI America™

Catalog No. D24625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24625G 5 g
D246225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24625G Supplier TCI America Supplier No. D24625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60956-23-2
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00079744
InChI Key LDCPXNOCWDGYIU-UHFFFAOYSA-N
Synonym 3,4-dibromotoluene, benzene, 1,2-dibromo-4-methyl, 3,4 dibromo toluene, benzene,1,2-dibromo-4-methyl, 3,4-dibromtoluol, 3,4 dibromotoluene, pubchem3930, acmc-1b64t, ksc493e6j
PubChem CID 34679
IUPAC Name 1,2-dibromo-4-methylbenzene
SMILES CC1=CC(=C(C=C1)Br)Br

Specifications

Melting Point -10°C
Color Yellow
Boiling Point 241°C
Quantity 5 g
Formula Weight 249.93
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4-Dibromotoluene