missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dibromotoluene, 98+%

Catalog No. AAA1335106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1335106 5 g
AAA1335114 25 g
AAA1335122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1335106 Supplier Thermo Scientific Chemicals Supplier No. A1335106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60956-23-2 | C7H6Br2 | 249.933 g/mol

It can be used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used in organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 60956-23-2
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00079744
InChI Key LDCPXNOCWDGYIU-UHFFFAOYSA-N
Synonym 3,4-dibromotoluene, benzene, 1,2-dibromo-4-methyl, 3,4 dibromo toluene, benzene,1,2-dibromo-4-methyl, 3,4-dibromtoluol, 3,4 dibromotoluene, pubchem3930, acmc-1b64t, ksc493e6j
PubChem CID 34679
IUPAC Name 1,2-dibromo-4-methylbenzene
SMILES CC1=CC(=C(C=C1)Br)Br

Specifications

Melting Point -10°C
Density 1.85
Boiling Point 251°C
Flash Point 116°C (241°F)
Refractive Index 1.6
Quantity 5 g
Beilstein 1931706
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 249.94
Percent Purity ≥98%
Chemical Name or Material 3,4-Dibromotoluene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only