missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dibromomaleimide 98.0+%, TCI America™
Supplier: TCI America D39105G
Specifications
| 3,4-Dibromomaleimide | |
| 228°C | |
| MFCD00185696 | |
| 1759 | |
| C1(=C(C(=O)NC1=O)Br)Br | |
| 254.865 | |
| 254.87 | |
| Crystalline Powder |
| 1122-10-7 | |
| C4HBr2NO2 | |
| 5 g | |
| BIKSKRPHKQWJCW-UHFFFAOYSA-N | |
| 3,4-dibromopyrrole-2,5-dione | |
| 14279 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 1122-10-7 | |
| 254.865 | |
| BIKSKRPHKQWJCW-UHFFFAOYSA-N | |
| 3,4-dibromopyrrole-2,5-dione |
| C4HBr2NO2 | |
| MFCD00185696 | |
| 14279 | |
| C1(=C(C(=O)NC1=O)Br)Br |
Safety and Handling
RTECSNumber : ON5100000
TSCA : No