accessibility menu, dialog, popup

UserName

CAS RN 1122-10-7

IUPAC Name: 3,4-dibromo-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 3,4-dibromo-1H-pyrrole-2,5-dione
Molecular Weight (g/mol): 254.87
Molecular Formula: C4HBr2NO2
InChi Key: BIKSKRPHKQWJCW-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C(=O)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results